Assay Details
Gene Symbol: gpank1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105703.2 | NP_001099173.2 | 2 - 3 | 332 | 80 | |
NM_001327760.1 | NP_001314689.1 | 2 - 3 | 316 | 80 | ||
NM_001327761.1 | NP_001314690.1 | 2 - 3 | 327 | 80 | ||
GenBank mRNA | BC151976.1 | - | 2 - 3 | 333 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
