Assay Details
Gene Symbol: vdac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001404.1 | NP_001001404.1 | 6 - 7 | 618 | 64 | |
GenBank mRNA | AY398429.1 | - | 6 - 7 | 618 | 64 | |
BC067647.1 | - | 6 - 7 | 580 | 64 | ||
BC160655.1 | - | 6 - 7 | 581 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
