Assay Details
Gene Symbol: ino80c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002108.2 | NP_001002108.2 | 3 - 4 | 533 | 152 | |
GenBank mRNA | BC071494.1 | - | 3 - 4 | 524 | 152 | |
EH554982.1 | - | 3 - 4 | 571 | 152 | ||
EH565174.1 | - | 2 - 3 | 373 | 152 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
