Assay Details
Gene Symbol: ino80c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002108.2 | NP_001002108.2 | 4 - 5 | 607 | 105 | |
GenBank mRNA | BC071494.1 | - | 4 - 5 | 598 | 105 | |
EH554982.1 | - | 4 - 5 | 645 | 105 | ||
EH565174.1 | - | 1 - 2 | 299 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
