Assay Details
Gene Symbol: rgcc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002190.1 | NP_001002190.1 | 2 - 3 | 305 | 102 | |
GenBank mRNA | BC074041.1 | - | 2 - 3 | 305 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
