Assay Details
Gene Symbol: eny2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002427.2 | NP_001002427.1 | 4 - 5 | 339 | 70 | |
GenBank mRNA | BC076140.1 | - | 4 - 5 | 288 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
