Assay Details
Gene Symbol: ndufa12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033103.2 | NP_001028275.1 | 3 - 4 | 334 | 75 | |
GenBank mRNA | BC100047.1 | - | 3 - 4 | 270 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
