Assay Details
Gene Symbol: taf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037431.1 | NP_001032508.1 | 2 - 3 | 675 | 95 | |
GenBank mRNA | BC067651.1 | - | 2 - 3 | 638 | 95 | |
BC083477.1 | - | 2 - 3 | 659 | 95 | ||
BC107956.1 | - | 2 - 3 | 675 | 95 | ||
BC129310.1 | - | 2 - 3 | 676 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
