Assay Details
Gene Symbol: taf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037431.1 | NP_001032508.1 | 8 - 9 | 1702 | 90 | |
GenBank mRNA | BC067651.1 | - | 8 - 9 | 1665 | 90 | |
BC107956.1 | - | 8 - 9 | 1702 | 90 | ||
BC129310.1 | - | 8 - 9 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
