Assay Details
Gene Symbol: taf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037431.1 | NP_001032508.1 | 9 - 10 | 1879 | 71 | |
GenBank mRNA | BC067651.1 | - | 9 - 10 | 1842 | 71 | |
BC107956.1 | - | 9 - 10 | 1879 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
