Assay Details
Gene Symbol: nenf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037704.2 | NP_001032793.2 | 1 - 2 | 443 | 111 | |
GenBank mRNA | AB178827.1 | - | 1 - 2 | 111 | ||
BC124089.1 | - | 1 - 2 | 195 | 111 | ||
BC162854.1 | - | 1 - 2 | 185 | 111 | ||
BC162880.1 | - | 1 - 2 | 183 | 111 | ||
EH553755.1 | - | 1 - 2 | 289 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
