Assay Details
Gene Symbol: chrna6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042684.2 | NP_001036149.1 | 2 - 3 | 363 | 67 | |
GenBank mRNA | BC171403.1 | - | 2 - 3 | 314 | 67 | |
BC171405.1 | - | 2 - 3 | 314 | 67 | ||
DQ822507.1 | - | 2 - 3 | 366 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
