Assay Details
Gene Symbol: pfdn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045289.1 | NP_001038754.1 | 1 - 2 | 75 | 68 | |
GenBank mRNA | BC074083.1 | - | 1 - 2 | 70 | 68 | |
BC095052.1 | - | 1 - 2 | 80 | 68 | ||
BC116551.1 | - | 1 - 2 | 75 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
