Assay Details
Gene Symbol: pfdn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045289.1 | NP_001038754.1 | 3 - 4 | 286 | 89 | |
GenBank mRNA | BC074083.1 | - | 3 - 4 | 281 | 89 | |
BC095052.1 | - | 3 - 4 | 291 | 89 | ||
BC116551.1 | - | 3 - 4 | 286 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
