Assay Details
Gene Symbol: cox5b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045357.1 | NP_001038822.1 | 3 - 4 | 323 | 72 | |
GenBank mRNA | BC122442.1 | - | 3 - 4 | 323 | 72 | |
BC153399.1 | - | 696 | 72 | |||
BC165491.1 | - | 3 - 4 | 300 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
