Assay Details
Gene Symbol: rnd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045364.1 | NP_001038829.1 | 1 - 2 | 219 | 81 | |
XM_017354589.2 | XP_017210078.1 | 1 - 2 | 364 | 81 | ||
GenBank mRNA | BC122188.1 | - | 1 - 2 | 219 | 81 | |
BC165068.1 | - | 1 - 2 | 136 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
