Assay Details
Gene Symbol: rnd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045364.1 | NP_001038829.1 | 3 - 4 | 425 | 72 | |
GenBank mRNA | BC122188.1 | - | 3 - 4 | 425 | 72 | |
BC165068.1 | - | 3 - 4 | 342 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
