Assay Details
Gene Symbol: bag6l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328276.1 | NP_001315205.1 | 3 - 4 | 344 | 68 | |
XM_017352268.2 | XP_017207757.1 | 3 - 4 | 397 | 68 | ||
XM_017352270.2 | XP_017207759.1 | 3 - 4 | 353 | 68 | ||
XM_017352271.2 | XP_017207760.1 | 3 - 4 | 397 | 68 | ||
XM_017352272.2 | XP_017207761.1 | 3 - 4 | 397 | 68 | ||
XM_017352273.2 | XP_017207762.1 | 3 - 4 | 353 | 68 | ||
GenBank mRNA | BC122262.1 | - | 3 - 4 | 344 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
