Assay Details
Gene Symbol: psmb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045564.2 | NP_001039029.2 | 6 - 7 | 671 | 104 | |
XM_021468901.1 | XP_021324576.1 | 6 - 7 | 669 | 104 | ||
GenBank mRNA | AF155581.1 | - | 6 - 7 | 569 | 104 | |
BC135050.1 | - | 6 - 7 | 583 | 104 | ||
BC163781.1 | - | 6 - 7 | 632 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
