Assay Details
Gene Symbol: josd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076595.2 | NP_001070063.1 | 4 - 5 | 1060 | 70 | |
GenBank mRNA | BC124423.1 | - | 4 - 5 | 1063 | 70 | |
BC152079.1 | - | 4 - 5 | 1045 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
