Assay Details
Gene Symbol: rps16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076732.2 | NP_001070200.1 | 3 - 4 | 186 | 92 | |
GenBank mRNA | BC122271.1 | - | 3 - 4 | 188 | 92 | |
BC164282.1 | - | 2 - 3 | 188 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
