Assay Details
Gene Symbol: cfap100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076760.1 | NP_001070228.1 | 6 - 7 | 746 | 128 | |
GenBank mRNA | BC124258.1 | - | 6 - 7 | 746 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
