Assay Details
Gene Symbol: lrrc51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076768.1 | NP_001070236.1 | 1 - 2 | 282 | 87 | |
XM_009291957.3 | XP_009290232.1 | 1 - 2 | 297 | 87 | ||
GenBank mRNA | BC091531.1 | - | 1 - 2 | 269 | 87 | |
BC124442.1 | - | 1 - 2 | 282 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
