Assay Details
Gene Symbol: hhatlb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077390.2 | NP_001070858.2 | 10 - 11 | 1394 | 67 | |
GenBank mRNA | BC124817.1 | - | 10 - 11 | 1384 | 67 | |
EH602025.1 | - | 2 - 3 | 171 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
