Assay Details
Gene Symbol: papss2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077767.1 | NP_001071235.1 | 3 - 4 | 435 | 75 | |
GenBank mRNA | BC127397.1 | - | 3 - 4 | 435 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
