Assay Details
Gene Symbol: f2rl1.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079805.1 | NP_001073273.1 | 1 - 2 | 107 | 83 | |
GenBank mRNA | BC128828.1 | - | 1 - 2 | 107 | 83 | |
BC171395.1 | - | 1 - 2 | 97 | 83 | ||
BC171397.1 | - | 1 - 2 | 97 | 83 | ||
GQ325254.1 | - | 1 - 2 | 70 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
