Assay Details
Gene Symbol: ppap2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079978.1 | NP_001073447.1 | 2 - 3 | 598 | 89 | |
GenBank mRNA | BC127580.1 | - | 2 - 3 | 598 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
