Assay Details
Gene Symbol: ppap2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079978.1 | NP_001073447.1 | 3 - 4 | 885 | 71 | |
GenBank mRNA | BC127580.1 | - | 3 - 4 | 885 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
