Assay Details
Gene Symbol: ppap2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079978.1 | NP_001073447.1 | 5 - 6 | 1123 | 107 | |
GenBank mRNA | BC127580.1 | - | 5 - 6 | 1123 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
