Assay Details
Gene Symbol: znf395a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080054.1 | NP_001073523.1 | 9 - 10 | 1308 | 80 | |
XM_005169829.4 | XP_005169886.1 | 9 - 10 | 1372 | 80 | ||
GenBank mRNA | BC128852.1 | - | 9 - 10 | 1308 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
