Assay Details
Gene Symbol: cracr2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080081.1 | NP_001073550.1 | 3 - 4 | 370 | 72 | |
XM_009297718.3 | XP_009295993.1 | 3 - 4 | 72 | |||
GenBank mRNA | BC128877.1 | - | 3 - 4 | 370 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
