Assay Details
Gene Symbol: surf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080173.1 | NP_001073642.1 | 5 - 6 | 576 | 115 | |
XM_009301509.2 | XP_009299784.1 | 2 - 3 | 280 | 115 | ||
GenBank mRNA | BC129348.1 | - | 5 - 6 | 576 | 115 | |
BC164262.1 | - | 5 - 6 | 580 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
