Assay Details
Gene Symbol: prf1.6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089507.1 | NP_001082976.1 | 2 - 3 | 563 | 104 | |
GenBank mRNA | BC139691.1 | - | 2 - 3 | 563 | 104 | |
KP099720.1 | - | 1 - 2 | 523 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
