Assay Details
Gene Symbol: arv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113611.1 | NP_001107083.1 | 1 - 2 | 131 | 110 | |
GenBank mRNA | BC154468.1 | - | 1 - 2 | 131 | 110 | |
BC171683.1 | - | 1 - 2 | 126 | 110 | ||
BC171685.1 | - | 1 - 2 | 126 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
