Assay Details
Gene Symbol: mlec | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113618.1 | NP_001107090.1 | 5 - 6 | 999 | 151 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
