Assay Details
Gene Symbol: akirin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113800.2 | NP_001107272.1 | 2 - 3 | 508 | 88 | |
GenBank mRNA | BC158177.1 | - | 2 - 3 | 441 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
