Assay Details
Gene Symbol: ramp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114432.1 | NP_001107904.1 | 4 - 5 | 233 | 61 | |
XM_017354532.2 | XP_017210021.1 | 4 - 5 | 280 | 61 | ||
GenBank mRNA | BC154812.1 | - | 4 - 5 | 233 | 61 | |
KM655677.1 | - | 4 - 5 | 205 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
