Assay Details
Gene Symbol: c1qtnf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114462.1 | NP_001107934.1 | 1 - 2 | 205 | 79 | |
XM_017353855.2 | XP_017209344.1 | 1 - 2 | 631 | 79 | ||
GenBank mRNA | BC159269.1 | - | 1 - 2 | 205 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
