Assay Details
Gene Symbol: c1qtnf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114462.1 | NP_001107934.1 | 2 - 3 | 261 | 81 | |
GenBank mRNA | BC159269.1 | - | 2 - 3 | 261 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
