Assay Details
Gene Symbol: ccnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131025.4 | NP_571100.1 | 1 - 2 | 484 | 119 | |
GenBank mRNA | AF365874.1 | - | 1 - 2 | 422 | 119 | |
BC075743.1 | - | 1 - 2 | 441 | 119 | ||
BC152628.1 | - | 1 - 2 | 440 | 119 | ||
EB896015.1 | - | 1 - 2 | 469 | 119 | ||
EH579652.1 | - | 1 - 2 | 474 | 119 | ||
X87581.1 | - | 402 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
