Assay Details
Gene Symbol: npy | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131074.2 | NP_571149.1 | 3 - 4 | 324 | 145 | |
GenBank mRNA | BC162068.1 | - | 3 - 4 | 324 | 145 | |
BC162071.1 | - | 3 - 4 | 324 | 145 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
