Assay Details
Gene Symbol: id1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131245.2 | NP_571320.2 | 1 - 2 | 464 | 81 | |
GenBank mRNA | AF007414.1 | - | 1 - 2 | 457 | 81 | |
BC071286.1 | - | 1 - 2 | 458 | 81 | ||
EH448502.1 | - | 1 - 2 | 492 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
