Assay Details
Gene Symbol: myb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309822.1 | NP_001296751.1 | 11 - 12 | 1556 | 66 | |
NM_131266.2 | NP_571341.2 | 10 - 11 | 1403 | 66 | ||
GenBank mRNA | AF191559.1 | - | 10 - 11 | 1334 | 66 | |
BC059803.1 | - | 11 - 12 | 1411 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
