Assay Details
Gene Symbol: myb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309822.1 | NP_001296751.1 | 12 - 13 | 1672 | 64 | |
NM_131266.2 | NP_571341.2 | 11 - 12 | 1519 | 64 | ||
GenBank mRNA | AF191559.1 | - | 11 - 12 | 1450 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
