Assay Details
Gene Symbol: klf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131859.3 | NP_571934.3 | 3 - 4 | 571 | 93 | |
XM_017356404.2 | XP_017211893.1 | 3 - 4 | 680 | 93 | ||
GenBank mRNA | AF392993.1 | - | 3 - 4 | 578 | 93 | |
BC066635.1 | - | 3 - 4 | 542 | 93 | ||
CK025353.1 | - | 3 - 4 | 518 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
