Assay Details
Gene Symbol: cxadr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152948.1 | NP_694480.1 | 1 - 2 | 163 | 76 | |
GenBank mRNA | AF268197.1 | - | 1 - 2 | 163 | 76 | |
BC045286.1 | - | 1 - 2 | 220 | 76 | ||
BC164203.1 | - | 1 - 2 | 86 | 76 | ||
BC171370.1 | - | 1 - 2 | 98 | 76 | ||
BC171372.1 | - | 1 - 2 | 98 | 76 | ||
CU458753.1 | - | 1 - 2 | 93 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
