Assay Details
Gene Symbol: cxadr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152948.1 | NP_694480.1 | 3 - 4 | 538 | 87 | |
GenBank mRNA | AF268197.1 | - | 3 - 4 | 538 | 87 | |
CU458753.1 | - | 3 - 4 | 468 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
