Assay Details
Gene Symbol: cxadr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152948.1 | NP_694480.1 | 4 - 5 | 692 | 96 | |
GenBank mRNA | AF268197.1 | - | 4 - 5 | 692 | 96 | |
BC045286.1 | - | 4 - 5 | 749 | 96 | ||
BC164203.1 | - | 4 - 5 | 615 | 96 | ||
BC171370.1 | - | 4 - 5 | 627 | 96 | ||
BC171372.1 | - | 4 - 5 | 627 | 96 | ||
CU458753.1 | - | 4 - 5 | 622 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
