Assay Details
Gene Symbol: cxadr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152948.1 | NP_694480.1 | 5 - 6 | 821 | 97 | |
GenBank mRNA | AF268197.1 | - | 5 - 6 | 821 | 97 | |
BC045286.1 | - | 5 - 6 | 878 | 97 | ||
BC164203.1 | - | 5 - 6 | 744 | 97 | ||
BC171370.1 | - | 5 - 6 | 756 | 97 | ||
BC171372.1 | - | 5 - 6 | 756 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
