Assay Details
Gene Symbol: jpt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY394952.1 | - | 1 - 2 | 177 | 112 | |
BC045436.1 | - | 1 - 2 | 156 | 112 | ||
BC066731.1 | - | 1 - 2 | 82 | 112 | ||
BC155180.1 | - | 1 - 2 | 101 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
